missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse PDGFA Protein
Recombinant Protein
Brand: enQuireBio™ QP5462-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_032834.1 | |
PDGFA Protein | |
Research Use Only | |
PDGFA | |
Recombinant Protein | |
E. coli | |
MSIEEAVPAV CKTRTVIYEI PRSQVDPTSA NFLIWPPCVE VKRCTGCCNT SSVKCQPSRV HHRSVKVAKV EYVRKKPKLK EVQVRLEEHL ECACATSNLN PDHREEETGR RRESGKNRKR KRLKPT | |
Greater than 95% as determined by SDS-PAGE. |
18590 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
3T3 cell proliferation significantly induced by less than 10 ng/ml of Mouse PDGFA (activity of at least 1.0 x 10^5 units/ml). | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |