missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Prealbumin / Transthyretin / TTR / PALB Protein
A DNA sequence encoding the Mus musculus (Mouse) Prealbumin / Transthyretin / TTR / PALB, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6849-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_038725.1 | |
Prealbumin / Transthyretin / TTR / PALB Protein | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
22139 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
TTR | |
Recombinant Protein | |
E. coli | |
GPAGAGESKC PLMVKVLDAV RGSPAVDVAV KVFKKTSEGS WEPFASGKTA ESGELHGLTT DEKFVEGVYR VELDTKSYWK TLGISPFHEF ADVVFTANDS GHRHYTIAAL LSPYSYSTTA VVSNPQN |