missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse R-Spondin 1 Fc Chimera Protein, CF

Product Code. 30226360
Click to view available options
Quantity:
1 mg
100 μg
20 μg
50 μg
Unit Size:
100µg
1mg
20µg
50µg
This item is not returnable. View return policy

Product Code. 30226360

Brand: R&D Systems™ 11566RS01M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Measured by its ability to activate TCF reporter activity in HEK293 human embryonic kidney cells in the presence of Wnt-3a. The ED50 for this effect is 5.00-75.0 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 284654
Molecular Weight (g/mol) M.W.-observed: 57-63 kDa under reducing conditions., M.W.-theroretical: 48 kDa
Quantity 1 mg
Source Chinese Hamster Ovary cell line, CHO-derived mouse R-Spondin 1 protein Mouse RSPO1 (Ser21-Pro207) (N-terminus) Accession # NP_619624.2 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias Cristin 3, CRISTIN3, FLJ40906Roof plate-specific spondin-1, hRspo1, roof plate-specific spondin, RSPO, RSPO1, RSPONDIN, RSpondin 1, R-spondin homolog (Xenopus laevis), R-spondin1, R-spondin-1
Conjugate Unconjugated
Reconstitution Reconstitute the 20 μg size at 200 μg/mL in PBS. Reconstitute all other sizes at 500 μg/mL in PBS.
Protein R-Spondin 1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.