missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse R-Spondin 1 Fc Chimera Protein, CF
Click to view available options
Quantity:
1 mg
100 μg
20 μg
50 μg
Unit Size:
100µg
1mg
20µg
50µg
Description
Measured by its ability to activate TCF reporter activity in HEK293 human embryonic kidney cells in the presence of Wnt-3a. The ED50 for this effect is 5.00-75.0 ng/mL.
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 284654 |
| Molecular Weight (g/mol) | M.W.-observed: 57-63 kDa under reducing conditions., M.W.-theroretical: 48 kDa |
| Quantity | 1 mg |
| Source | Chinese Hamster Ovary cell line, CHO-derived mouse R-Spondin 1 protein Mouse RSPO1 (Ser21-Pro207) (N-terminus) Accession # NP_619624.2 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | Cristin 3, CRISTIN3, FLJ40906Roof plate-specific spondin-1, hRspo1, roof plate-specific spondin, RSPO, RSPO1, RSPONDIN, RSpondin 1, R-spondin homolog (Xenopus laevis), R-spondin1, R-spondin-1 |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction