missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse RANKL / OPGL / TNFSF11 / CD254 Protein

Product Code. p-7152095
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
500 μg
Unit Size:
100µg
10µg
1mg
2µg
500µg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15936049 2 μg 2µg
15916049 10 μg 10µg
15906049 100 μg 100µg
15946049 500 μg 500µg
15926049 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15936049 Supplier enQuireBio™ Supplier No. QP54022ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_035743.2
Gene ID (Entrez) 21943
Name RANKL / OPGL / TNFSF11 / CD254 Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol TNFSF11
Biological Activity Mouse RANKL / OPGL / TNFSF11 / CD254 induces TNF-alpha production in RAW cells (typical ED50 is 50-75 ng/ml). Activity was further confirmed by measuring Osteoclast formation (significanty different from control at Mouse RANKL / OPGL / TNFSF11 / CD254 concentrations greater than 50 ng/ml).
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag Untagged
Sequence MPAMMEGSWL DVAQRGKPEA QPFAHLTINA ASIPSGSHKV TLSSWYHDRG WAKISNMTLS NGKLRVNQDG FYYLYANICF RHHETSGSVP TDYLQLMVYV VKTSIKIPSS HNLMKGGSTK NWSGNSEFHF YSINVGGFFK LRAGEEISIQ VSNPSLLDPD QDATYFGAFK VQDID
Buffer 10 mM sodium phosphate and 20 mM sodium chloride, pH 7.5
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.