missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Ras-related protein Rab-10 Protein

A DNA sequence encoding the Mus musculus (Mouse) Ras-related protein Rab-10, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6565-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16034210

  • 3375.00 € / 1mg
Estimated Shipment: 03-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_057885.1
Ras-related protein Rab-10 Protein
1 mg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
19325
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Rab10
Recombinant Protein
E. coli
MAKKTYDLLF KLLLIGDSGV GKTCVLFRFS DDAFNTTFIS TIGIDFKIKT VELQGKKIKL QIWDTAGQER FHTITTSYYR GAMGIMLVYD ITNGKSFENI SKWLRNIDEH ANEDVERMLL GNKCDMDDKR VVPKGKGEQI AREHGIRFFE TSAKANINIE KAFLTLAEDI LRKTPVKEPN SENVDISSGG GVTGWKSKCC
Videos
SDS
Documents
Certificates
Special Offers

Special Offers