missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Reg3B Protein

Product Code. 18757897
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18757897 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18757897 Supplier R&D Systems™ Supplier No. 5110RG050
  • ONLINE EXCLUSIVE PRICE
    414.36 €
    368.55 € / 50µg
    Save 45.81 €
    11% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Reg3B Protein is derived from E. coli. The Recombinant Mouse Reg3B Protein has been validated for the following applications: Bioactivity.

Order Info

The product is shipped at ambient temperature. Upon receipt, store it immediately at the recommended storage temperatures.

TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 18489
Quantity 50 μg
Source E. coli-derived mouse Reg3B protein Glu27-Gly175,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias PAP1, Reg3B
Conjugate Unconjugated
Product Line R&D Systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.