missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse ROR1 Fc Chimera Protein

Product Code. 18777866
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18777866 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18777866 Supplier R&D Systems™ Supplier No. 9910RO050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse ROR1 Fc Chimera Protein is derived from NS0. The Recombinant Mouse ROR1 Fc Chimera Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Z139
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 4919
Molecular Weight (g/mol) 69 kDa
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse ROR1 protein, (N-terminus) Mouse ROR1 (Gln30-Glu403) Accession &Num; Q9Z139 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements 12 months from date of receipt, ≤ -20°C as supplied; 1 month, 2 to 8°C under sterile conditions after reconstitution; 3 months, ≤ -20°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Cross Reactivity ROR1
Species Mouse
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.