missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Roundabout homolog 1 Protein

Product Code. p-7147977
Click to view available options
Quantity:
100 μg
20 μg
50 μg
Unit Size:
100µg
20µg
50µg
This item is not returnable. View return policy

Product Code. 15902858

Brand: enQuireBio™ QP10099ma20ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) Roundabout homolog 1, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_062286.2
Gene ID (Entrez) 19876
Name Roundabout homolog 1 Protein
Quantity 20 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Robo1
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Mouse
Species Mammal
Protein Tag His
Sequence YRHRKKRNGL TSTYAGIRKV PSFTFTPTVT YQRGGEAVSS GGRPGLLNIS EPATQPWLAD TWPNTGNNHN DCSINCCTAG NGNSDSNLTT YSRPADCIAN YNNQLDNKQT NLMLPESTVY GDVDLSNKIN EMKTFNSPNL KDGRFVNPSG QPTPYATTQL IQANLSNNMN NGAGDSSEKH WKPPGQQKPE VAPIQYNIME QNKLNKDYRA NDTIPPTIPY NQSYDQNTGG SYNSSDRGSS TSGSQGHKKG ARTPKAPKQG GMNWADLLPP PPAHPPPHSN SEEYNMSVDE SYDQEMPCPV PPAPMYLQQD ELQEEEDERG PTPPVRGAAS SPAAVSYSHQ STATLTPSPQ EELQPMLQDC PEDLGHMPHP PDRRRQPVSP PPPPRPISPP HTYGYISGPL VSDMDTDAPE EEEDEADMEV AKMQTRRLLL RGLEQTPASS VGDLESSVTG SMINGWGSAS EEDNISSGRS SVSSSDGSFF TDADFAMS-Q AVAAAAEYAG LKVARRQMQD AAGRRHFHAS QCPRPTSPVS TDSNMSAVVI QKARPAKKQK HQPGHLRREA YADDLPPPPV PPPAIKSPTV QSKAQLEVRP VMVPKLASIE ARTDRSSDRK GGSYKGREAL DGRQVTDLRT NPSDPREAQE QPNDGKGRGT RQPKRDLPPA KTHLGQEDIL PYCRPTFPTS NNPRDPSSSS SMSSRGSGSR QREQANVGRR NMAEMQVLGG FERGDENNEE LEETES
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.