missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse S100A4 Protein

A cDNA sequence encoding the S100A4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13394-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15923429

  • 203.00 € / 20µg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

S100A4 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MARPLEEALD VIVSTFHKYS GKEGDKFKLN KTELKELLTR ELPSFLGKRT DEAAFQKVMS NLDSNRDNEV DFQEYCVFLS CIAMMCNEFF EGCPDKEPRK K
Greater than 90.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
His
S100A4 protein solution (1 mg/1ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 2mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers