missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse S100A6 Protein

A cDNA sequence encoding the S100A6 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13397-HIS-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15933439

  • 3595.00 € / 1mg
Estimated Shipment: 12-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

20200
Greater than 90% as determined by SDS-PAGE.
Research Use Only
S100a6
Mouse
His
The protein was lyophilized from a concentrated solution (1 mg/ml) containing no additives.
S100A6 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MACPLDQAIG LLVAIFHKYS GKEGDKHTLS KKELKELIQK ELTIGSKLQD AEIARLMDDL DRNKDQEVNF QEYVAFLGAL ALIYNEALK
Videos
SDS
Documents
Certificates
Special Offers

Special Offers