missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse S100A8 / CAGA Protein

A DNA sequence encoding the Mus musculus (Mouse) S100A8 / CAGA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6641-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024740

  • 735.00 € / 100µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_038678.1
S100A8 / CAGA Protein
Research Use Only
S100A8
Recombinant Protein
E. coli
PSELEKALSN LIDVYHNYSN IQGNHHALYK NDFKKMVTTE CPQFVQNINI ENLFRELDIN SDNAINFEEF LAMVIKVGVA SHKDSHKE
Greater than 90% as determined by SDS-PAGE.
20201
100 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers