missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse SERPINA6 Protein

Product Code. 15923639
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15923639

Brand: enQuireBio™ QP134592ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the SERPINA6 was constructed and used to recombinantly synthesize the protein.

Specifications

Name SERPINA6 Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Mouse
Species Human (HEK293)
Protein Tag His
Sequence VTDEDSSSHR DLAPTNVDFA FNLYKRLVAL NSDKNTLISP VSISMALAML SLSTRGSTQY LENLGFNMSK MSEAEIHQGF QYLNSLLQQS DTGLEMNMGN VMFLLQNLKL KDSFLADTKH YYESEALTIP SKDWTKAGEQ INNHVKNKTQ GKIEHVVSDL DSSATLILIN YIFLKGIWKL PFSPENTREE DFYVNETSTV KVPMMVQSGN ISYFRDSAIP CQMVQMNYVG NGTTFIILPD QGQMDTVVAA LNRDTIDRWG KLMIPRQMNL YIPKFSMSDT YDLQDVLADV GIKDLFTNQS DFADTTKDTP LTLTVLHKAM LQLDEGNVLP AATNGPPVHL PSESFTLKYN RPFIFLAFDK YTWSSLMMSQ VMNPA HHHHH HHHH
Buffer SERPINA6 was filtered (0.4microm) and lyophilized in phosphate buffered saline pH 8.0.
Purity or Quality Grade Greater than 95.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.