missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Serum Amyloid A2 Protein

Product Code. 18767617
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18767617 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18767617 Supplier R&D Systems™ Supplier No. 8478SA050
  • ONLINE EXCLUSIVE PRICE
    494.16 €
    441.00 € / 50µg
    Save 53.16 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to induce TNF-alpha secretion by J774A.1 mouse reticulum cell sarcoma macrophage cells. The ED50 for this effect is 1.5-7.5 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05367
For Use With (Application) Bioactivity
Gene ID (Entrez) 6289
Molecular Weight (g/mol) Observed M.W.: 11kDa, reducing conditions, Theoretical M.W.: 12kDa
Quantity 50 μg
Source E. coli-derived mouse Serum Amyloid A2 protein Gly20-Tyr122
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias SAA, SAA2, serum amyloid A2
Common Name SAA2
Gene Symbol Saa2
Biological Activity Measured by its ability to induce TNF-alpha secretion by J774A.1 mouse reticulum cell sarcoma macrophage cells. The ED50 for this effect is typically 1.5-7.5μg/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System E. coli-derived Gly20-Tyr122
Product Line R&D Systems
Purity or Quality Grade >85%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.