missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse SET domain-containing protein 1
A DNA sequence encoding the Mus musculus (Mouse) SET domain-containing protein 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP10045-ba-20ug
Additional Details : Weight : 0.01000kg
Specifications
NP_663457.2 | |
SET domain-containing protein 1 | |
Research Use Only | |
Setd4 | |
Recombinant Protein | |
Baculovirus | |
MQRRRGRTER ARKRRRRSSG SRAVNESYRS EFIELRKWLK ERKFEDTDLV PASFPGTGRG LMSKASLQEG QVMISLPESC LLTTDTVIRS SLGPYIKKWK PPVSPLLALC TFLVSEKHAG CRSLWKSYLD ILPKSYTCPV CLEPEVVDLL PSPLKAKAEE QRARVQDLFT SARGFFSTLQ PLFAEPVDSV FSYRAFLWAW CTVNTRAVYL RSRRQECLSA EPDTCALAPF LDLLNHSPHV QVKAAFNEKT RCYEIRTASR CRKHQEVFIC YGPHDNQRLL LEYGFVSVRN PHACVPVSAD MLVKFLPAAD KQLHRKITIL KDHGFTGNLT FGWDGPSWRL LTALKLLCLE AERFTSWKKV LLGEVISDTN EKTSLGVAQK ICSDVIEETH AVLRKVSDMK EGTVSLRNQL SLVEALRMEE LRILQASAEI LSGLLAPFS | |
Greater than 80% as determined by SDS-PAGE. |
224440 | |
20 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |