missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Siglec-H Fc Chimera Protein

Product Code. 18789786
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18789786 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18789786 Supplier R&D Systems™ Supplier No. 10264SH050
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Measured by its binding ability in a functional ELISA.

When Recombinant Mouse Siglec-H Fc Chimera is immobilized at 2 μg/mL (100 μL/well), the concentration of Recombinant Mouse Siglec-3 Fc Chimera that produces 50% of the optimal binding response is 5 to 20 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_848821.2
For Use With (Application) Bioactivity
Gene ID (Entrez) 233274
Molecular Weight (g/mol) 55 kDa
Name 6430529G09Rik, SiglecH, Siglec-H, Siglech sialic acid binding Ig-like lectin H
Quantity 50 μg
Source

Mouse myeloma cell line, NS0-derived mouse Siglec-H protein

Mouse Siglec-H (Asn19-Gly269) Accession # NP_848821.2IEGRMDPMouse IgG2a (Glu98-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Mouse
Reconstitution Reconstitute at 500 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Product Line R&D Systems
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.