missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Survivin / BIRC5 / API4 Protein

A DNA sequence encoding the Mus musculus (Mouse) Survivin / BIRC5 / API4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5720-ye-500ug

 View more versions of this product

Product Code. 15967259

  • 2100.00 € / 500µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001012273.1
Survivin / BIRC5 / API4 Protein
Research Use Only
BIRC5
Recombinant Protein
Yeast
MGAPALPQIW QLYLKNYRIA TFKNWPFLED CACTPERMAE AGFIHCPTEN EPDLAQCFFC FKELEGWEPD DNPIEEHRKH SPGCAFLTVK KQMEELTVSE FLKLDRQRAK NKIAKETNNK QKEFEETAKT TRQSIEQLAA
Greater than 90% as determined by SDS-PAGE.
11799
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales