missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Sushi repeat-containing protein SRPX2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Sushi repeat-containing protein SRPX2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7826-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001077364.2 | |
Sushi repeat-containing protein SRPX2 Protein | |
Research Use Only | |
Srpx2 | |
Recombinant Protein | |
E. coli | |
WYAGSGYSPD ESYNEVYAEE VPAARARALD YRVPRWCYTL NIQDGEATCY SPRGGNYHSS LGTRCELSCD RGFRLIGRKS VQCLPSRRWS GTAYCRQIRC HTLPFITSGT YTCTNGMLLD SRCDYSCSSG YHLEGDRSRI CMEDGRWSGG EPVCVDIDPP KIRCPHSREK MAEPEKLTAR VYWDPPLVKD SADGTITRVT LRGPEPGSHF PEGEHVIRYT AYDRAYNRAS CKFIVKVQVR RCPILKPPQH GYLTCSSAGD NYGAICEYHC DGGYERQGTP SRVCQSSRQW SGTPPVCTPM KINVNVNSAA GLLDQFYEKQ RLLIVSAPDP SNRYYKMQIS MLQQSTCGLD LRHVTIIELV GQPPQEVGRI REQQLSAGII EELRQFQRLT RSYFNMVLID KQGIDRERYM EPVTPEEIFT FIDDYLLSNE ELARRVEQRD LCE | |
Greater than 80% as determined by SDS-PAGE. |
68792 | |
10 μg | |
Not determined. | |
Full Length of Mature Protein | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |