missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Sushi repeat-containing protein SRPX2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Sushi repeat-containing protein SRPX2, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_001077364.2 |
---|---|
Gene ID (Entrez) | 68792 |
Name | Sushi repeat-containing protein SRPX2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16011031
|
enQuireBio™
QP7826-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16031031
|
enQuireBio™
QP7826-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16021031
|
enQuireBio™
QP7826-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041031
|
enQuireBio™
QP7826-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16051031
|
enQuireBio™
QP7826-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061031
|
enQuireBio™
QP7826-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001077364.2 | |
Sushi repeat-containing protein SRPX2 Protein | |
Not determined. | |
Full Length of Mature Protein | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
68792 | |
Research Use Only | |
Srpx2 | |
Recombinant Protein | |
E. coli | |
WYAGSGYSPD ESYNEVYAEE VPAARARALD YRVPRWCYTL NIQDGEATCY SPRGGNYHSS LGTRCELSCD RGFRLIGRKS VQCLPSRRWS GTAYCRQIRC HTLPFITSGT YTCTNGMLLD SRCDYSCSSG YHLEGDRSRI CMEDGRWSGG EPVCVDIDPP KIRCPHSREK MAEPEKLTAR VYWDPPLVKD SADGTITRVT LRGPEPGSHF PEGEHVIRYT AYDRAYNRAS CKFIVKVQVR RCPILKPPQH GYLTCSSAGD NYGAICEYHC DGGYERQGTP SRVCQSSRQW SGTPPVCTPM KINVNVNSAA GLLDQFYEKQ RLLIVSAPDP SNRYYKMQIS MLQQSTCGLD LRHVTIIELV GQPPQEVGRI REQQLSAGII EELRQFQRLT RSYFNMVLID KQGIDRERYM EPVTPEEIFT FIDDYLLSNE ELARRVEQRD LCE | |
Greater than 80% as determined by SDS-PAGE. |