missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Thrombospondin-2 Protein

A DNA sequence encoding the Mus musculus (Mouse) Thrombospondin-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6786-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16035840

  • 735.00 € / 100µg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_035711.2
Thrombospondin-2 Protein
Research Use Only
Thbs2
Recombinant Protein
E. coli
GDHVKDTSFD LFSISNINRK TIGAKQFRGP DPGVPAYRFV RFDYIPPVNT DDLNRIVKLA RRKEGFFLTA QLKQDRKSRG TLLVLEGPGT SQRQFEIVSN GPGDTLDLNY WVEGNQHTNF LEDVGLADSQ WKNVTVQVAS DTYSLYVGCD LIDSVTLEEP FYEQLEVDRS RMYVAKGASR ESHFRGLLQN VHLVFADSVE DILSKKGCQH SQGA
Greater than 90% as determined by SDS-PAGE.
21826
100 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers