missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TNFR2 Protein
A cDNA sequence encoding the TNFR2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10886-1mg
Additional Details : Weight : 0.01000kg
Specifications
21938 | |
Greater than 97.0% as determined by SDS-PAGE. | |
Research Use Only | |
Tnfrsf1b | |
Recombinant Protein | |
E. coli | |
MPAQVAFTPY APEPGSTCRL REYYDQTAQM CCSKCSPGQH AKVFCTKTSD TVCDSCEDST YTQLWNWVPE CLSCGSRCSS DQVETQACTR EQNRICTCRP GWYCALSKQE GCRLCAPLRK CRPGFGVARP GTETSDVVCK PCAPGTFSNT TSSTDICRPH QICNVVAIPG NASMDAVCTS TSPT |
TNFR2 Protein | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by its ability to inhibit the TNF-? mediated cytotoxicity in the L-929 cells is less than 0.2microg/ml, corresponding to a specific activity of > 5000IU/mg in the presence of 0.25ng/mL of rHuTNF-a. | |
Mouse | |
Untagged | |
TNFR2 protein was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |