missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Transcriptional repressor CTCFL Protein

A DNA sequence encoding the Mus musculus (Mouse) Transcriptional repressor CTCFL, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5884-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15958449

  • 3260.00 € / 1mg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001074856.1
Transcriptional repressor CTCFL Protein
Research Use Only
Ctcfl
Recombinant Protein
Yeast
MAAAEVPVPS GYFTQIKEQK LKPGDLEEEK EEDGVQRVEA QEGVVKEVEA ENSCLLLEAR APVESDRRIL TLQTVHLESQ DVHLQGLGWL SVPHSEELSG TVPEAEGILQ LPSVLWLDPE PQLSLQHCVT VSIPEELYPP EELQRIHFHL LRENVLMAEE NPELTPDLDE STALKKPEED EKDQLPPQGE TDKREERLLL LEMKPKEGKD DEIVLTISHL SLEEQQDPPA ANQTSVPGAK AAKPKRRRQT KGKPQSFQCD TCPFTSSKLS TFNRHIKIHS NERPHLCHLC LKAFRTVTLL RNHVNTHTGT RPHKCRDCDM AFVTSGELVR HRRYKHTYEK PFKCSLCKYA SVEASKMKRH IRSHTGERPF QCCQCAYASR DSYKLKRHMR THSGEKPYEC PTCHVRFTQS GTMKIHIAQK HGENVPKYEC PHCATIIARK SDLRVHLRNL HSQSPEEMKC RYCPAGFHER YALIQHQRTH KNEKKFKCKQ CDYACKQERC LKAHMRMHTG EKPFSCLACN KHFRQKQLLT VHLRKYHDPN FVPNLHLCLK CDKRFSRWSN LQRHRKKCDP EHETLAPNKD RRPVTRTQAS EGEAGHKEGE PQCPGEQALG HQGEAAGSQS PDHGLTCEII FNMMDK
Greater than 90% as determined by SDS-PAGE.
664799
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers