missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Transmembrane protease serine 4 Protein

A DNA sequence encoding the Mus musculus (Mouse) Transmembrane protease serine 4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7925-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16011631

  • 735.00 € / 100µg
Estimated Shipment: 24-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_663378.1
Transmembrane protease serine 4 Protein
Research Use Only
Tmprss4
Recombinant Protein
E. coli
KVILDKYYFI CGSPLTFIQR GQLCDGHLDC ASGEDEEHCV KDFPEKPGVA VRLSKDRSTL QVLDAATGTW ASVCFDNFTE ALAKTACRQM GYDSQPAFRA VEIRPDQNLP VAQVTGNSQE LQVQNGSRSC LSGSLVSLRC LDCGKSLKTP RVVGGVEAPV DSWPWQVSIQ YNKQHVCGGS ILDPHWILTA AHCFRKYLDV SSWKVRAGSN ILGNSPSLPV AKIFIAEPNP LYPKEKDIAL VKLQMPLTFS GSVRPICLPF SDEVLVPATP VWVIGWGFTE ENGGKMSDML LQASVQVIDS TRCNAEDAYE GEVTAEMLCA GTPQGGKDTC QGDSGGPLMY HSDKWQVVGI VSWGHGCGGP STPGVYTKVT AYLNWIYNVR KSEM
Greater than 90% as determined by SDS-PAGE.
214523
100 μg
Not determined.
Extracellular Domain
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers