missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse VAMP2 Protein
A cDNA sequence encoding the VAMP2 was constructed and used to recombinantly synthesize the protein.
131.00 € - 1740.00 €
Specifications
Name | VAMP2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15985109
|
enQuireBio™
QP13916-1UG |
1 μg |
131.00 €
1µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15905119
|
enQuireBio™
QP13916-5UG |
5 μg |
203.00 €
5µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15995109
|
enQuireBio™
QP13916-50UG |
50 μg |
1740.00 €
50µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
VAMP2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
VAMP2 protein solution (1 mg/ml) containing Phosphate Buffered Saline (pH 7.4), 1mM EDTA, 0.1mM PMSF and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSATAA TVPPAAPAGE GGPPAPPPNL TSNRRLQQTQ AQVDEVVDIM RVNVDKVLER DQKLSELDDR ADALQAGASQ FETSAAKLKR KYWWKNLK | |
Greater than 90.0% as determined by SDS-PAGE. |