missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse VEGF-164 Isoform Protein

Product Code. 15985699
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
500 μg
Unit Size:
100µg
10µg
1mg
2µg
500µg
This item is not returnable. View return policy

Product Code. 15985699

Brand: enQuireBio™ QP52142ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_001273986.1
Name VEGF-164 Isoform Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol VEGFA
Biological Activity HUVEC proliferation was significantly induced by less than 10 ng/ml, or activity of at least 1.0 x 10^5 units/mg. Typical ED50 values of new lots are generally lower, around 5 ng/ml.
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag Untagged
Sequence MAPTTEGEQK SHEVIKFMDV YQRSYCRPIE TLVDIFQEYP DEIEYIFKPS CVPLMRCAGC CNDEALECVP TSESNITMQI MRIKPHQSQH IGEMSFLQHS RCECRPKKDR TKPENHCEPC SERRKHLFVQ DPQTCKCSCK NTDSRCKARQ LELNERTCRC DKPRR
Buffer Sterile filtered (0.22 um) solution containing 0.1% TFA
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.