missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse VEGF-164 Isoform Protein
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
500 μg
Unit Size:
100µg
10µg
1mg
2µg
500µg
Specifications
Specifications
| Accession Number | NP_001273986.1 |
| Name | VEGF-164 Isoform Protein |
| Quantity | 100 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
| Gene Symbol | VEGFA |
| Biological Activity | HUVEC proliferation was significantly induced by less than 10 ng/ml, or activity of at least 1.0 x 10^5 units/mg. Typical ED50 values of new lots are generally lower, around 5 ng/ml. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Mouse |
| Species | E. coli |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction