missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Multi Species PKA-RII alpha Protein

Product Code. 15982389
Click to view available options
Quantity:
1 μg
100 μg
3 μg
Unit Size:
100µg
1µg
3µg
This item is not returnable. View return policy

Product Code. 15982389

Brand: enQuireBio™ QP130631ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the PKA-RII alpha was constructed and used to recombinantly synthesize the protein.

Specifications

Name Multi Species PKA-RII alpha Protein
Quantity 1 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Multi Species
Species E. coli
Protein Tag Untagged
Sequence MSHIQIPPGL TELLQGYTVE VLRQQPPDLV EFAVEYFTRL REARAPASVL PAATPRQSLG HPPPEPGPDR VADAKGDSES EEDEDLEVPV PSRFNRRVSV CAETYNPDEE EEDTDPRVIH PKTDEQRCRL QEACKDILLF KNLDQEQLSQ VLDAMFERIV KADEHVIDQG DDGDNFYVIE RGTYDILVTK DNQTRSVGQY DNRGSFGELA LMYNTPRAAT IVATSEGSLW GLDRVTFRRI IVKNNAKKRK MFESFIESVP LLKSLEVSER MKIVDVIGEK IYKDGERIIT QGEKADSFYI IESGEVSILI RSRTKSNKDG GNQEVEIARC HKGQYFGELA LVTNKPRAAS AYAVGDVKCL VMDVQAFERL LGPCMDIMKR NISHYEEQLV KMFGSSVDLG NLGQ
Buffer PKA regulatory subunit-II alpha is supplied in 20mM MES, 150mM NaCl, 2mM EDTA, 2mM EGTA, 1mM BME and 50% glycerol.
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.