missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mycoplasma Chaperone protein DnaK Protein

A DNA sequence encoding the Mycoplasma pneumoniae (strain ATCC 29342 / M129) Chaperone protein DnaK, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6942-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16036800

  • 940.00 € / 100µg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_110122.1
Mycoplasma Chaperone protein DnaK Protein
Research Use Only
dnaK
Recombinant Protein
E. coli
QGERPMARDN KSLGRFNLGG IQPAPKGKPQ IEITFSLDAN GILNVKAKDL TTQKENSITI SDNGNLSEEE IQKMIRDAEA NKERDNVIRE RIELRNEGES IVSTIKEILQ SPEAKDFPKE EKEKLDKITG GIDAAIKAND YTKLKAEIEN FKKWREEMAK KYNPNGDQGQ PAQ
Greater than 90% as determined by SDS-PAGE.
876838
100 μg
Not determined.
Partial (see sequence information for more details).
Mycoplasma
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers