missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mycoplasma High affinity transport system protein p37 Protein

A DNA sequence encoding the Mycoplasma hyorhinis High affinity transport system protein p37, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7079-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16037310

  • 3590.00 € / 1mg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Mycoplasma High affinity transport system protein p37 Protein
Research Use Only
p37
Recombinant Protein
E. coli
CSNTGVVKQE DVSVSQGQWD KSITFGVSEA WLNKKKGGEK VNKEVINTFL ENFKKEFNKL KNANDKTKNF DDVDFKVTPI QDFTVLLNNL STDNPELDFG INASGKLVEF LKNNPGIITP ALETTTNSFV FDKEKDKFYV DGTDSDPLVK IAKEINKIFV ETPYASWTDE NHKWNGNVYQ SVYDPTVQAN FYRGMIWIKG NDETLAKIKK AWNDKDWNTF RNFGILHGKD NSFSKFKLEE TILKNHFQNK FTTLNEDRSA HPNAYKQKSA DTLGTLDDFH IAFSEEGSFA WTHNKSATKP FETKANEKME ALIVTNPIPY DVGVFRKSVN QLEQNLIVQT FINLAKNKQD TYGPLLGYNG YKKIDNFQKE IVEVYEKAIK
Greater than 90% as determined by SDS-PAGE.
1 mg
Not determined.
Full Length
Mycoplasma
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers