missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mycoplasma Lon protease Protein
A DNA sequence encoding the Mycoplasma pneumoniae (strain ATCC 29342 / M129) Lon protease, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6966-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_110020.1 | |
Mycoplasma Lon protease Protein | |
Research Use Only | |
lon | |
Recombinant Protein | |
E. coli | |
MPAVKKPQIL VVRNQVIFPY NGFELDVGRE RSKKLIKALK NLKTKRLVLV TQKNSDQLNP EFDDIYHCGT LCDIDEIIEV PSEDGKTADY KIKGKGLQRV AITSFSDADL TKYDHHFLNS TLTENKALDK LLERIFPDKE DFAEILDSLN SFLELQELKK LSKVPKDIKR YDIITFKLAS LIFKDITLQQ AILEENDIEK RLQKII | |
Greater than 90% as determined by SDS-PAGE. |
877195 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mycoplasma | |
His-SUMO | |
Tris-based buffer, 50% glycerol |