missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mycoplasma Lon protease Protein

A DNA sequence encoding the Mycoplasma pneumoniae (strain ATCC 29342 / M129) Lon protease, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6966-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16046910

  • 2260.00 € / 500µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_110020.1
Mycoplasma Lon protease Protein
Research Use Only
lon
Recombinant Protein
E. coli
MPAVKKPQIL VVRNQVIFPY NGFELDVGRE RSKKLIKALK NLKTKRLVLV TQKNSDQLNP EFDDIYHCGT LCDIDEIIEV PSEDGKTADY KIKGKGLQRV AITSFSDADL TKYDHHFLNS TLTENKALDK LLERIFPDKE DFAEILDSLN SFLELQELKK LSKVPKDIKR YDIITFKLAS LIFKDITLQQ AILEENDIEK RLQKII
Greater than 90% as determined by SDS-PAGE.
877195
500 μg
Not determined.
Partial (see sequence information for more details).
Mycoplasma
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers