missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mycoplasma Putative proline iminopeptidase Protein
A DNA sequence encoding the Mycoplasma pneumoniae (strain ATCC 29342 / M129) Putative proline iminopeptidase, was expressed in the hosts and tags indicated.
421.00 € - 3590.00 €
Specifications
Accession Number | NP_109710.1 |
---|---|
Gene ID (Entrez) | 876978 |
Name | Mycoplasma Putative proline iminopeptidase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16096890
|
enQuireBio™
QP6964-10UG |
10 μg |
421.00 €
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16036900
|
enQuireBio™
QP6964-50UG |
50 μg |
605.00 €
50µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16086890
|
enQuireBio™
QP6964-100UG |
100 μg |
1025.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16016900
|
enQuireBio™
QP6964-200UG |
200 μg |
1690.00 €
200µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16026900
|
enQuireBio™
QP6964-500UG |
500 μg |
2260.00 €
500µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16006900
|
enQuireBio™
QP6964-1MG |
1 mg |
3590.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_109710.1 | |
Mycoplasma Putative proline iminopeptidase Protein | |
Not determined. | |
Full Length | |
Mycoplasma | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
876978 | |
Research Use Only | |
Pip | |
Recombinant Protein | |
E. coli | |
MNTSPKQSGY LKVGNGHEVY FWTAGNPQGK SALYVHGGPG SGTDAGCLKY FDLDTTYVIL LDQRGCGQSK AVNPLLHNTT QDLVGDLEAL RQHLKLERWT LFGGSWGSTL ALVYAITHPQ VVEQVFLRAL FLGREQDWAE MLLGLGKLFY PYEHQTLLKA IPQACRTDFT KFTNYFYEVL QGNDSALKTQ LANAWVKWEN TLLSPISYVK DEKAEDANFT FKLALLECHY AKHHSFLKPN FILENVAVLK DKPVHLIHGR FDLVCPLSQA LELKRALPTL NLYVTNNAGH SGSDPNNLTT IKHLLKTQL | |
Greater than 80% as determined by SDS-PAGE. |