missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other AHSG Protein
A cDNA sequence encoding the AHSG was constructed and used to recombinantly synthesize the protein.
131.00 € - 3480.00 €
Specifications
Name | AHSG Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15945768
|
enQuireBio™
QP10982-10UG |
10 μg |
131.00 €
10µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965768
|
enQuireBio™
QP10982-50UG |
50 μg |
203.00 €
50µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955768
|
enQuireBio™
QP10982-1MG |
1 mg |
3480.00 €
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
AHSG Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Filtered (0.4microm) and lyophilized from 0.5 mg/ml in PBS pH 7.4. |
Research Use Only | |
Recombinant Protein | |
Non-species Specific | |
MKSLVLLLCL AQLWGCHSAP HGPGLIYRQP NCDDPETEEA ALVAIDYINQ NLPWGYKHTL NQIDEVKVWP QQPSGELFEI EIDTLETTCH VLDPTPVARC SVRQLKEHAV EGDCDFQLLK LDGKFSVVYA KCDSSPDSAE DVRKVCQDCP LLAPLNDTRV VHAAKAALAA FNAQNNGSNF QLEEISRAQL VPLPPSTYVE FTVSGTDCVA KEATEAAKCN LLAEKQYGFC KATLSEKLGG AEVAVTCTVF QTQPVTSQPQ PEGANEAVPT PVVDPDAPPS PPLGAPGLPP AGSPPDSHVL LAAPPGHQLH RAHYDLRHTF MGVVSLGSPS GEVSHPRKTR TVVQPSVGAA AGPVVPPCPG RIRHFKV | |
Greater than 95% as determined by SDS-PAGE. |