missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Alarelin Protein
Recombinant Protein
131.00 € - 570.00 €
Specifications
Name | Alarelin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15965818
|
enQuireBio™
QP10997-2MG |
2 mg |
131.00 €
2mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955818
|
enQuireBio™
QP10997-10MG |
10 mg |
203.00 €
10mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945818
|
enQuireBio™
QP10997-100MG |
100 mg |
570.00 €
100mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Alarelin Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
5-oxo-pro-His-Trp-Ser-Tyr-D-Ala-Leu-Arg-Pro-Nhet x CH3COOH | |
Greater than 98.0% as determined by Analysis by RP-HPLC. |
Research Use Only | |
Recombinant Protein | |
Untagged | |
The Alarelin was lyophilized with no additives. |