missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Angiostatin K1-4 Protein
A cDNA sequence encoding the Angiostatin K1-4 was constructed and used to recombinantly synthesize the protein.
131.00 € - 6960.00 €
Specifications
Name | Angiostatin K1-4 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | Human Angiostatin Kringles 1-4 significantly inhibits basic-FGF induced endothelial cell proliferation and migration at concentration ranging from 300nM-1.0 uM. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15934128
|
enQuireBio™
QP10521-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15914128
|
enQuireBio™
QP10521-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924128
|
enQuireBio™
QP10521-1MG |
1 mg |
6960.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Angiostatin K1-4 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Non-species Specific | |
Lyophilized from a (1 mg/ml) solution in containing 20mM Hepes buffer pH 8.2 and 20mM NaCl. |
Research Use Only | |
Human Angiostatin Kringles 1-4 significantly inhibits basic-FGF induced endothelial cell proliferation and migration at concentration ranging from 300nM-1.0 uM. | |
Other | |
Untagged | |
Greater than 98.0% as determined by SDS-PAGE. |