missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Avidin Protein

Product Code. p-7148326
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 15954138

Brand: enQuireBio™ QP105255mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Avidin was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 396260
Name Avidin Protein
Quantity 5 mg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol AVD
Biological Activity 16 units/mg protein, 1 unit binds 1microg biotin.
Product Type Recombinant Protein
Cross Reactivity Other
Species Non-species Specific
Protein Tag Untagged
Sequence MVHATSPLLL LLLLSLALVA PGLSARKCSL TGKWTNDLGS NMTIGAVNSR GEFTGTYITA VTATSNEIKE SPLHGTQNTI NKRTQPTFGF TVNWKFSEST TVFTGQCFID RNGKEVLKTM WLLRSSVNDI GDDWKATRVG INIFTRLRTQ KE
Purity or Quality Grade Greater than 92.0%.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.