missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Batroxobin Protein
Click to view available options
Quantity:
1 mg
10 μg
50 μg
Unit Size:
10µg
1mg
50µg
Specifications
Specifications
| Name | Batroxobin Protein |
| Quantity | 10 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Other |
| Species | E. coli |
| Protein Tag | Untagged |
| Sequence | MVLIRVIANL LILQVSYAQK SSELVIGGDE CDINEHPFLA FMYYSPRYFC GMTLINQEWV LTAAHCNRRF MRIHLGKHAG SVANYDEVVR YPKEKFICPN KKKNVITDKD IMLIRLDRPV KNSEHIAPLS LPSNPPSVGS VCRIMGWGAI TTSEDTYPDV PHCANINLFN NTVCREAYNG LPAKTLCAGV LQGGIDTCGG DSGGPLICNG QFQGILSWGS DPCAEPRKPA FYTKVFDYLP WIQSIIAGNK TATCP |
| Buffer | The Batroxobin protein was lyophilized from a concentrated (1 mg/ml) solution with no additives. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction