missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Bivalirudin Protein

A cDNA sequence encoding the Bivalirudin was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11165-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15976348

  • 131.00 € / 1mg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Bivalirudin Protein
Research Use Only
Recombinant Protein
E. coli
The protein (1 mg/ml) was lyophilized with 0.5 mg Manntiol and sodium hydroxide 50microg pH 5.5.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers