missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other CYP2D6 Protein

A cDNA sequence encoding the CYP2D6 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11583-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15917718

  • 203.00 € / 20µg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CYP2D6 Protein
Research Use Only
Recombinant Protein
Baculovirus
MGLEALVPLA VIVAIFLLLV DLMHRRQRWA ARYPPGPLPL PGLGNLLHVD FQNTPYCFDQ LRRRFGDVFS LQLAWTPVVV LNGLAAVREA LVTHGEDTAD RPPVPITQIL GFGPRSQGVF LARYGPAWRE QRRFSVSTLR NLGLGKKSLE QWVTEEAACL CAAFANHSGR PFRPNGLLDK AVSNVIASLT CGRRFEYDDP RFLRLLDLAQ EGLKEESGFL REVLNAVPVL LHIPALAGKV LRFQKAFLTQ LDELLTEHRM TWDPAQPPRD LTEAFLAEME KAKGNPESSF NDENLRIVVA DLFSAGMVTT STTLAWGLLL MILHPDVQRR VQQEIDDVIG QVRRPEMGDQ AHMPYTTAVI HEVQRFGDIV PLGVTHMTSR DIEVQGFRIP KGTTLITNLS SVLKDEAVWE KPFRFHPEHF LDAQGHFVKP EAFLPFSAGR RACLGEPLAR MELFLFFTSL LQHFSFSVPT GQPRPSHHGV FAFLVSPSPY ELCAVPR
Greater than 40% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
LKM-1 is supplied in 50mM Potassium phosphate buffer pH 7.5, 15% glycerol, 1mM DTT and 1% Triton X-100.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers