missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other CYP2D6 Protein
A cDNA sequence encoding the CYP2D6 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11583-5ug
Additional Details : Weight : 0.01000kg
Specifications
CYP2D6 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MGLEALVPLA VIVAIFLLLV DLMHRRQRWA ARYPPGPLPL PGLGNLLHVD FQNTPYCFDQ LRRRFGDVFS LQLAWTPVVV LNGLAAVREA LVTHGEDTAD RPPVPITQIL GFGPRSQGVF LARYGPAWRE QRRFSVSTLR NLGLGKKSLE QWVTEEAACL CAAFANHSGR PFRPNGLLDK AVSNVIASLT CGRRFEYDDP RFLRLLDLAQ EGLKEESGFL REVLNAVPVL LHIPALAGKV LRFQKAFLTQ LDELLTEHRM TWDPAQPPRD LTEAFLAEME KAKGNPESSF NDENLRIVVA DLFSAGMVTT STTLAWGLLL MILHPDVQRR VQQEIDDVIG QVRRPEMGDQ AHMPYTTAVI HEVQRFGDIV PLGVTHMTSR DIEVQGFRIP KGTTLITNLS SVLKDEAVWE KPFRFHPEHF LDAQGHFVKP EAFLPFSAGR RACLGEPLAR MELFLFFTSL LQHFSFSVPT GQPRPSHHGV FAFLVSPSPY ELCAVPR | |
Greater than 40% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
LKM-1 is supplied in 50mM Potassium phosphate buffer pH 7.5, 15% glycerol, 1mM DTT and 1% Triton X-100. |