missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Cys-Protein-G Protein

Product Code. 15947728
Click to view available options
Quantity:
1 mg
10 mg
100 mg
Unit Size:
100mg
10mg
1mg
This item is not returnable. View return policy

Product Code. 15947728

Brand: enQuireBio™ QP115871mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Cys-Protein-G was constructed and used to recombinantly synthesize the protein.

Specifications

Name Cys-Protein-G Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Other
Species E. coli
Protein Tag Untagged
Buffer Lyophilized without any additives.
Purity or Quality Grade Greater than 95.0% as determined by SDS-PAGE and HPLC analyses.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.