missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other DnaK Mycobacterium Tuberculosis Protein
A cDNA sequence encoding the DnaK Mycobacterium Tuberculosis was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11686-2ug
Additional Details : Weight : 0.01000kg
Specifications
908588 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MARAVGIDLG TTNSVVSVLE GGDPVVVANS EGSRTTPSTV AFARNGEVLV GQPAKNQAVT NVDRTIRSVK RHMGSDWSIE IDGKKYTAQE ISARVLMKLK RDAEAYLGED ITDAVITTPA YFNDAQRQAT KEAGQIAGLN VLRIVNEPTA AALAYGLDKG EREQTILVFD LGGGTFDVSL LEIGEGVVEV RATSGDNHLG GDDWDDRIVN WLVDKFKGTS GIDLTKDKMA MQRLREAAEK AKIELSSSQS TSVNLPYITV DSDKNPLFLD EQLIRAEFQR ITQDLLDRTR QPFQSVVKDA GISVSEIDHV VLVGGSTRMP AVTDLVKELT GGKEPNKGVN PDEVVAVGAA LQAGVLKGEV KDVLLLDVTP LSLGIETKGG VMTKLIERNT TIPTKRSETF TTADDNQPSV QIQVYQGERE IASHNKLLGS FELTGIPPAP RGVPQIEVTF DIDANGIVHV TAKDKGTGKE NTIKIQEGSG LSKEEIDRMV KDAEAHAEED RKRREEADVR NQAETLVYQT EKFVKEQRET ENGSRVPEDT LNKVEAAVAE AKTALGGTDI SAIKSAMEKL GQDSQALGQA IYEATQAASK VGGEASAPGG SNSTDDVVDA EVVDDERESK | |
Greater than 95.0% as determined by SDS-PAGE. |
DnaK Mycobacterium Tuberculosis Protein | |
Research Use Only | |
dnaK | |
Other | |
Untagged | |
The DnaK protein was lyophilized from a concentrated (1 mg/ml) solution containing 10mM Na-phosphate buffer pH 7.4, 130mM NaCl and 2.5mM KCl. |