missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other GHRP 6 Protein
A cDNA sequence encoding the GHRP 6 was constructed and used to recombinantly synthesize the protein.
131.00 € - 570.00 €
Specifications
Name | GHRP 6 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15988958
|
enQuireBio™
QP11969-5MG |
5 mg |
131.00 €
5mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15978958
|
enQuireBio™
QP11969-25MG |
25 mg |
203.00 €
25mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15968958
|
enQuireBio™
QP11969-100MG |
100 mg |
570.00 €
100mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
GHRP 6 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
The GHRP-6 peptide was lyophilized with no additives. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
His-D-Trp-Ala-Trp-D-Phe-Lys-NH2 | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |