missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Gliadin Gamma 18.6kDa Wheat Protein

A cDNA sequence encoding the Gliadin Gamma 18.6kDa Wheat was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11984-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15929008

  • 120.00 € / 5µg
Estimated Shipment: 19-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Gliadin Gamma 18.6kDa Wheat Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Greater than 94% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
Gliadin is supplied in 20mM HEPES buffer pH 8.0, 200mM NaCl, and 20% glycerol.
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers