missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Hexarelin Protein

Product Code. 15929758
Change view
Click to view available options
Quantity:
20 mg
100 mg
500 mg
Unit Size:
100mg
20mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15929758 500 mg 500mg
15919758 20 mg 20mg
15909758 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15929758 Supplier enQuireBio™ Supplier No. QP12225500mg
  • ONLINE EXCLUSIVE PRICE
    4966.50 €
    3820.84 € / 500mg
    Save 1145.66 €
    23% Off
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A cDNA sequence encoding the Hexarelin was constructed and used to recombinantly synthesize the protein.

Specifications

Name Hexarelin Protein
Quantity 500 mg
Regulatory Status RUO
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Other
Species E. coli
Protein Tag Untagged
Buffer The Hexarelin peptide was lyophilized with no additives.
Purity or Quality Grade Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.