missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Hirudin Protein
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
Specifications
Specifications
| Name | Hirudin Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Biological Activity | The specific activity was found to be >14,000ATU/mg. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Other |
| Species | P. pastoris |
| Protein Tag | Untagged |
| Buffer | Each mg of protein was lyophilized from a sterile solution containing 20mM PBS pH 7 and 2% mannitol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction