missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other ldhA Protein

A cDNA sequence encoding the ldhA was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12548-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15940779

  • 203.00 € / 20µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

946315
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMKLAVY STKQYDKKYL QQVNESFGFE LEFFDFLLTE KTAKTANGCE AVCIFVNDDG SRPVLEELKK HGVKYIALRC AGFNNVDLDA AKELGLKVVR VPAYDPEAVA EHAIGMMMTL NRRIHRAYQR TRDANFSLEG LTGFTMYGKT AGVIGTGKIG VAMLRILKGF GMRLLAFDPY PSAAALELGV EYVDLPTLFS ESDVISLHCP LTPENYHLLN EAAFEQMKNG VMIVNTSRGA LIDSQAAIEA LKNQKIGSLG MDVYENERDL FFEDKSNDVI QDDVFRRLSA CHNVLFTGHQ AFLTAEALTS ISQTTLQNLS NLEKGETCPN ELV
Greater than 95% as determined by SDS-PAGE.
ldhA Protein
Research Use Only
ldhA
Other
His
The ldhA solution (1 mg/ml) contains 20mM Tris-HCl buffer, pH 8.0, 10% glycerol and 100mM NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers