missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other MIF T. Vaginalis Protein

A cDNA sequence encoding the MIF T. Vaginalis was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12697-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15961259

  • 203.00 € / 10µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MIF T. Vaginalis Protein
Research Use Only
Recombinant Protein
E. coli
MPALVIKTNA KFTEEEKSKA TEELGNIVSK VLGKPISYVM VTLEDGVAVR FGGSDEKAAF MSLMSIGGLN RAVNKRASAA LTKWFTDHGF QGDRIYIVFN PKSAEDWGFN GDTFAHHHHH H
Greater than 95.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
MIF T. Vaginalis filtered (0.4 microm) and lyophilized from 0.5 mg/ml solution in 20 mM Tris buffer and 50 mM NaCl, pH 7.5.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers