missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other MoeX Protein
A cDNA sequence encoding the MoeX was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12716-100ug
Additional Details : Weight : 0.01000kg
Spezifikation
MoeX Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
PBS. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Protein is >95% pure as determined by 10% PAGE (coomassie staining). |