missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Myelin Basic Protein
A cDNA sequence encoding the Myelin Basic Protein was constructed and used to recombinantly synthesize the protein.
131.00 € - 5220.00 €
Specifications
Gene ID (Entrez) | 7008 |
---|---|
Name | Myelin Basic Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | TEF |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15931139
|
enQuireBio™
QP12658-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15951139
|
enQuireBio™
QP12658-HIS-EC-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15911139
|
enQuireBio™
QP12658-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15941139
|
enQuireBio™
QP12658-HIS-EC-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15921139
|
enQuireBio™
QP12658-1MG |
1 mg |
5220.00 €
1mg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
7008 | |
Research Use Only | |
TEF | |
Other | |
His | |
The MBP solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 10% glycerol. |
Myelin Basic Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGNHAGK RELNAEKAST NSETNRGESE KKRNLGELSR TTSEDNEVFG EADANQNNGT SSQDTAVTDS KRTADPKNAW QDAHPADPGS RPHLIRLFSR DAPGREDNTF KDRPSESDEL QTIQEDSAAT SESLDVMASQ KRPSQRHGSK YLATASTMDH ARHGFLPRHR DTGILDSIGR FFGGDRGAPK RGSGKVSSEE | |
Greater than 85% as determined by SDS-PAGE. |